Product Details

SNP ID
rs200094928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17516267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGCGGCTGAGGACTACGCCA[A/C]GAAGCTGAGACAGGTGAGCCCCGAG
Phenotype
MIM: 609967 MIM: 604951
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM129C PubMed Links

Gene Details

Gene
FAM129C
Gene Name
family with sequence similarity 129 member C
There are no transcripts associated with this gene.

Gene
PGLS
Gene Name
6-phosphogluconolactonase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012088.2 1033 Missense Mutation AAG,ACG K128T NP_036220.1
XM_011527897.2 1033 Missense Mutation AAG,ACG K40T XP_011526199.1

View Full Product Details