Product Details

SNP ID
rs200161870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45499246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTTTCTGCGGAGCCAGAGGTGGC[C/G]GCACTGGCACGCCAGGCTGAGGACG
Phenotype
MIM: 603183 MIM: 601703
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPM1N PubMed Links

Gene Details

Gene
PPM1N
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080401.1 774 Silent Mutation GCC,GCG A258A NP_001073870.1
XM_011526474.2 774 Silent Mutation GCC,GCG A258A XP_011524776.1
Gene
RTN2
Gene Name
reticulon 2
There are no transcripts associated with this gene.

Gene
VASP
Gene Name
vasodilator-stimulated phosphoprotein
There are no transcripts associated with this gene.

View Full Product Details