Product Details

SNP ID
rs199500012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13912530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGGCTGTGTGTCCCTGCAGGC[A/G]GAGCTAAATGAGGTCCTTGGAGAGG
Phenotype
MIM: 610055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf57 PubMed Links

Gene Details

Gene
C19orf57
Gene Name
chromosome 19 open reading frame 57
There are no transcripts associated with this gene.

Gene
CC2D1A
Gene Name
coiled-coil and C2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017721.4 614 Silent Mutation GCA,GCG A105A NP_060191.3
XM_005259972.2 614 Silent Mutation GCA,GCG A105A XP_005260029.1
XM_005259973.2 614 Silent Mutation GCA,GCG A105A XP_005260030.1
XM_005259974.2 614 Silent Mutation GCA,GCG A105A XP_005260031.1
XM_005259975.2 614 Silent Mutation GCA,GCG A105A XP_005260032.1

View Full Product Details