Product Details

SNP ID
rs200097552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55227556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGGTGGGCGGTGGCAAAGGC[A/G]GCCATGCCTGGCGGGAGAGGGGTGG
Phenotype
MIM: 610875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6804 PubMed Links

Gene Details

Gene
MIR6804
Gene Name
microRNA 6804
There are no transcripts associated with this gene.

Gene
PPP6R1
Gene Name
protein phosphatase 6 regulatory subunit 1
There are no transcripts associated with this gene.

Gene
TMEM86B
Gene Name
transmembrane protein 86B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173804.4 602 Silent Mutation NP_776165.3
XM_017026554.1 602 Silent Mutation XP_016882043.1

View Full Product Details