Product Details

SNP ID
rs200201061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42300060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCACCTGCTCTGCTGTGTGTCC[A/G]TGGTTGTTGGTGCCCACCCAGACCA
Phenotype
MIM: 612082 MIM: 603074
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIC PubMed Links

Gene Details

Gene
CIC
Gene Name
capicua transcriptional repressor
There are no transcripts associated with this gene.

Gene
PAFAH1B3
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145939.1 653 Silent Mutation CAC,CAT H106H NP_001139411.1
NM_001145940.1 653 Silent Mutation CAC,CAT H106H NP_001139412.1
NM_002573.3 653 Silent Mutation CAC,CAT H106H NP_002564.1
XM_017026846.1 653 Silent Mutation CAC,CAT H106H XP_016882335.1
XM_017026847.1 653 Silent Mutation CAC,CAT H106H XP_016882336.1
XM_017026848.1 653 Silent Mutation CAC,CAT H106H XP_016882337.1
Gene
PRR19
Gene Name
proline rich 19
There are no transcripts associated with this gene.

View Full Product Details