Product Details

SNP ID
rs199855150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57232577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGCGCCTGTATAACTATTTCC[A/G]TGATGCACGCCGGGTGTACCTGATT
Phenotype
MIM: 603495 MIM: 604668
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKC PubMed Links

Gene Details

Gene
AURKC
Gene Name
aurora kinase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015878.1 380 Missense Mutation CAT,CGT H111R NP_001015878.1
NM_001015879.1 380 Missense Mutation CAT,CGT H92R NP_001015879.1
NM_003160.2 380 Missense Mutation CAT,CGT H77R NP_003151.2
Gene
ZNF264
Gene Name
zinc finger protein 264
There are no transcripts associated with this gene.

Gene
ZNF805
Gene Name
zinc finger protein 805
There are no transcripts associated with this gene.

View Full Product Details