Product Details

SNP ID
rs200470776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:362227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCCAGGCCTGGGTTGATCACA[C/T]TTTTCTGGTGATGCTTTTGGGGGTG
Phenotype
MIM: 609503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THEG PubMed Links

Gene Details

Gene
THEG
Gene Name
theg spermatid protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016585.4 1858 Silent Mutation AAA,AAG K371K NP_057669.1
NM_199202.2 1858 Silent Mutation AAA,AAG K347K NP_954672.1
XM_011528049.2 1858 Silent Mutation AAA,AAG K439K XP_011526351.1
XM_011528050.2 1858 Silent Mutation AAA,AAG K415K XP_011526352.1
XM_011528051.2 1858 Silent Mutation AAA,AAG K371K XP_011526353.1
XM_011528052.2 1858 Intron XP_011526354.1

View Full Product Details