Product Details

SNP ID
rs199646908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2290636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCGCCGCGCACCTCGGCCGGGG[G/T]GGCGCAGGCCGGCAGCCGCCCGTCG
Phenotype
MIM: 609792
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf35 PubMed Links

Gene Details

Gene
C19orf35
Gene Name
chromosome 19 open reading frame 35
There are no transcripts associated with this gene.

Gene
LINGO3
Gene Name
leucine rich repeat and Ig domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101391.1 1269 Missense Mutation ACC,CCC T381P NP_001094861.1

View Full Product Details