Product Details

SNP ID
rs200031770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15914629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGTCTGGCCACACAGTTGGGTT[C/G]TAATGGATCCCGATAATATTGATGA
Phenotype
MIM: 611517
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP4F11 PubMed Links

Gene Details

Gene
CYP4F11
Gene Name
cytochrome P450 family 4 subfamily F member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128932.1 1745 Silent Mutation TAC,TAG Y429* NP_001122404.1
NM_021187.3 1745 Silent Mutation TAC,TAG Y429* NP_067010.3

View Full Product Details