Product Details
- SNP ID
-
rs200032167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:34759567 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCGGTATGGGTCCTCTTATGTC[A/G]GATGAAAGTGGAGCGATGAGTAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF599
PubMed Links
Gene Details
- Gene
- ZNF599
- Gene Name
- zinc finger protein 599
View Full Product Details