Product Details

SNP ID
rs200135869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9296247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACATTTATAGGGTTTCTCTCCA[A/G]TATGTGTCCTCCCATGTACAGTGAG
Phenotype
MIM: 609571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF699 PubMed Links

Gene Details

Gene
ZNF699
Gene Name
zinc finger protein 699
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198535.2 1527 Missense Mutation NP_940937.1

View Full Product Details