Product Details

SNP ID
rs200275281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39504079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGTCCCCCATAGTGGTGTGC[C/G]GAGCAGGCTGCAGCCCTGAGCATGG
Phenotype
MIM: 602768
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016941.3 719 Missense Mutation CGA,GGA R221G NP_058637.1
NM_203486.2 719 Missense Mutation CGA,GGA R221G NP_982353.1

View Full Product Details