Product Details

SNP ID
rs200364034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19277837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAGCCGATGTTCTCCACAGTCA[A/G]CTTGAACTCCTTGTACTCTGAGTAG
Phenotype
MIM: 607992 MIM: 606563
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUGP1 PubMed Links

Gene Details

Gene
SUGP1
Gene Name
SURP and G-patch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172231.3 Intron NP_757386.2
XM_005260002.2 Intron XP_005260059.1
XM_011528155.2 Intron XP_011526457.1
Gene
TM6SF2
Gene Name
transmembrane 6 superfamily member 2
There are no transcripts associated with this gene.

View Full Product Details