Product Details

SNP ID
rs199545548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5914729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCTGGATCTGGAGGAGTTCCTT[C/T]GGGCGCTGCGGGTGAGCCCCCACCT
Phenotype
MIM: 114212 MIM: 603327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPS PubMed Links

Gene Details

Gene
CAPS
Gene Name
calcyphosine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004058.4 938 Missense Mutation CGG,TGG R170W NP_004049.2
NM_080590.3 938 Missense Mutation CGG,TGG R170W NP_542157.2
Gene
RANBP3
Gene Name
RAN binding protein 3
There are no transcripts associated with this gene.

Gene
VMAC
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details