Product Details

SNP ID
rs199618668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21809199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGTAGAATTTCTCTCCAGTGC[A/G]AATTATCTTATGGGTAGTAAGGATT
Phenotype
MIM: 603972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF43 PubMed Links

Gene Details

Gene
ZNF43
Gene Name
zinc finger protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256648.1 6666 Intron NP_001243577.1
NM_001256649.1 6666 Missense Mutation CGC,TGC R274C NP_001243578.1
NM_001256650.1 6666 Intron NP_001243579.1
NM_001256651.1 6666 Intron NP_001243580.1
NM_001256653.1 6666 Intron NP_001243582.1
NM_001256654.1 6666 Missense Mutation CGC,TGC R215C NP_001243583.1
NM_003423.3 6666 Missense Mutation CGC,TGC R280C NP_003414.2
XM_011528257.2 6666 Intron XP_011526559.1
XM_011528259.2 6666 Missense Mutation CGC,TGC R274C XP_011526561.1
XM_017027207.1 6666 Intron XP_016882696.1
XM_017027208.1 6666 Intron XP_016882697.1
XM_017027209.1 6666 Intron XP_016882698.1
XM_017027210.1 6666 Intron XP_016882699.1
XM_017027211.1 6666 Missense Mutation CGC,TGC R274C XP_016882700.1
XM_017027212.1 6666 Intron XP_016882701.1
XM_017027213.1 6666 Intron XP_016882702.1
XM_017027214.1 6666 Intron XP_016882703.1
XM_017027215.1 6666 Intron XP_016882704.1
XM_017027216.1 6666 Intron XP_016882705.1

View Full Product Details