Product Details

SNP ID
rs199652968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56221585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGGATTATGCAATCACTGAGAA[A/G]TAGCTTCTGGATGTGTTTTCTGGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN5A PubMed Links

Gene Details

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 1830 Missense Mutation NP_001308990.1
NM_001322062.1 1830 Intron NP_001308991.1
NM_001322064.1 1830 Missense Mutation NP_001308993.1
NM_001322065.1 1830 Missense Mutation NP_001308994.1
NM_001322066.1 1830 Missense Mutation NP_001308995.1
NM_001322067.1 1830 Missense Mutation NP_001308996.1
NM_001322068.1 1830 Missense Mutation NP_001308997.1
NM_001322069.1 1830 Missense Mutation NP_001308998.1
NM_001322070.1 1830 Missense Mutation NP_001308999.1
NM_001322072.1 1830 Missense Mutation NP_001309001.1
NM_001322073.1 1830 Missense Mutation NP_001309002.1
NM_001322074.1 1830 Missense Mutation NP_001309003.1
NM_001322075.1 1830 Missense Mutation NP_001309004.1
NM_001322076.1 1830 Missense Mutation NP_001309005.1
NM_001322077.1 1830 Missense Mutation NP_001309006.1
NM_001322078.1 1830 Missense Mutation NP_001309007.1
NM_024303.2 1830 Intron NP_077279.1
XM_005259254.1 1830 Intron XP_005259311.1
XM_006723377.3 1830 Missense Mutation XP_006723440.1
XM_011527308.1 1830 Intron XP_011525610.1
XM_017027298.1 1830 Missense Mutation XP_016882787.1
XM_017027299.1 1830 Missense Mutation XP_016882788.1

View Full Product Details