Product Details

SNP ID
rs199829487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17826830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCGGCTTCCGCTCCACAGCAT[A/G]TCCAGCTGGGGGCCCAGGGCGCTGA
Phenotype
MIM: 146738 MIM: 600173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSL3 PubMed Links

Gene Details

Gene
INSL3
Gene Name
insulin like 3
There are no transcripts associated with this gene.

Gene
JAK3
Gene Name
Janus kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000215.3 3388 Silent Mutation GAC,GAT D1096D NP_000206.2
XM_005259896.3 3388 Silent Mutation GAC,GAT D1139D XP_005259953.1
XM_011527990.1 3388 Intron XP_011526292.1
XM_011527991.2 3388 Intron XP_011526293.1

View Full Product Details