Product Details

SNP ID
rs200074258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52155114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACATTTGTATGGCTTCTCTCCAG[C/T]GTGATTTCTTTGATGGTACACCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF836 PubMed Links

Gene Details

Gene
ZNF836
Gene Name
zinc finger protein 836
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102657.1 2978 Missense Mutation ACT,GCT T857A NP_001096127.1
XM_011526558.2 2978 Missense Mutation ACT,GCT T857A XP_011524860.1
XM_011526559.2 2978 Missense Mutation ACT,GCT T857A XP_011524861.1
XM_017026389.1 2978 Intron XP_016881878.1

View Full Product Details