Product Details
- SNP ID
-
rs200282023
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:38213705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACTCGATGCACTGCCAGCGGTAG[G/T]TCCGCACGGCTGCCGTCATGTTCAC
- Phenotype
-
MIM: 601670
MIM: 616655
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DPF1
PubMed Links
Gene Details
- Gene
- DPF1
- Gene Name
- double PHD fingers 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135155.2 |
1007 |
Missense Mutation |
AAC,ACC |
N344T |
NP_001128627.1 |
NM_001135156.2 |
1007 |
Missense Mutation |
AAC,ACC |
N262T |
NP_001128628.1 |
NM_001289978.1 |
1007 |
Missense Mutation |
AAC,ACC |
N344T |
NP_001276907.1 |
NM_004647.3 |
1007 |
Missense Mutation |
AAC,ACC |
N300T |
NP_004638.2 |
XM_005259289.2 |
1007 |
Missense Mutation |
AAC,ACC |
N327T |
XP_005259346.1 |
XM_005259290.3 |
1007 |
Missense Mutation |
AAC,ACC |
N318T |
XP_005259347.1 |
XM_005259292.4 |
1007 |
Missense Mutation |
AAC,ACC |
N299T |
XP_005259349.1 |
XM_006723407.2 |
1007 |
Missense Mutation |
AAC,ACC |
N318T |
XP_006723470.1 |
XM_006723408.3 |
1007 |
Missense Mutation |
AAC,ACC |
N316T |
XP_006723471.1 |
XM_006723409.2 |
1007 |
Missense Mutation |
AAC,ACC |
N162T |
XP_006723472.1 |
XM_006723410.2 |
1007 |
Missense Mutation |
AAC,ACC |
N162T |
XP_006723473.1 |
XM_011527356.1 |
1007 |
Missense Mutation |
AAC,ACC |
N327T |
XP_011525658.1 |
XM_011527357.2 |
1007 |
Missense Mutation |
AAC,ACC |
N299T |
XP_011525659.1 |
XM_011527358.2 |
1007 |
Missense Mutation |
AAC,ACC |
N262T |
XP_011525660.1 |
- Gene
- SIPA1L3
- Gene Name
- signal induced proliferation associated 1 like 3
There are no transcripts associated with this gene.
View Full Product Details