Product Details

SNP ID
rs199653947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16185687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGCAATGGGAACGAGCAAAAA[C/G]AACGAGGAGGAGAAGCGGCGCGGCC
Phenotype
MIM: 614554
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM32A PubMed Links

Gene Details

Gene
FAM32A
Gene Name
family with sequence similarity 32 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014077.3 397 Intron NP_054796.1
XM_011527898.1 397 Missense Mutation CAA,GAA Q128E XP_011526200.1

View Full Product Details