Product Details

SNP ID
rs199689586
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6468369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACAAATCCAGTTCTTCTCCAGA[C/T]CCCAAGAAGCTGCTGTCCAGAGCTT
Phenotype
MIM: 609737 MIM: 613634 MIM: 608746
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CRB3 PubMed Links
Additional Information
For this assay, SNP(s) [rs62107586] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CRB3
Gene Name
crumbs 3, cell polarity complex component
There are no transcripts associated with this gene.

Gene
DENND1C
Gene Name
DENN domain containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290331.1 1698 Silent Mutation GGA,GGG G508G NP_001277260.1
NM_024898.3 1698 Silent Mutation GGA,GGG G552G NP_079174.2
XM_006722905.2 1698 Silent Mutation GGA,GGG G466G XP_006722968.2
XM_006722906.3 1698 Silent Mutation GGA,GGG G508G XP_006722969.1
XM_011528318.2 1698 Silent Mutation GGA,GGG G508G XP_011526620.1
Gene
SLC25A23
Gene Name
solute carrier family 25 member 23
There are no transcripts associated with this gene.

View Full Product Details