Product Details

SNP ID
rs199725672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1986602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGCGCCCGTGGTGGGCGACTC[A/G]TGTGTCACCTTGCGCAGGCCTTTGG
Phenotype
MIM: 608531 MIM: 602214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD2 PubMed Links

Gene Details

Gene
BTBD2
Gene Name
BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017797.3 830 Silent Mutation NP_060267.2
XM_005259593.1 830 Silent Mutation XP_005259650.1
XM_011528127.1 830 Silent Mutation XP_011526429.1
Gene
CSNK1G2
Gene Name
casein kinase 1 gamma 2
There are no transcripts associated with this gene.

View Full Product Details