Product Details

SNP ID
rs199793891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38905302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGGCGGAGCGTAGGGGCCACA[C/T]GGCGCTGCACCTGGCCTGCCGTGTG
Phenotype
MIM: 604495 MIM: 612804 MIM: 604480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCER2 PubMed Links

Gene Details

Gene
CCER2
Gene Name
coiled-coil glutamate rich protein 2
There are no transcripts associated with this gene.

Gene
NFKBIB
Gene Name
NFKB inhibitor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243116.1 425 Missense Mutation ACG,ATG T43M NP_001230045.1
NM_002503.4 425 Missense Mutation ACG,ATG T129M NP_002494.2
XM_006723226.2 425 Missense Mutation ACG,ATG T97M XP_006723289.2
XM_006723227.3 425 Missense Mutation ACG,ATG T124M XP_006723290.3
Gene
SARS2
Gene Name
seryl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
SIRT2
Gene Name
sirtuin 2
There are no transcripts associated with this gene.

View Full Product Details