Product Details

SNP ID
rs199902802
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43479379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGTGGCCAGGCTGGGCTTACCC[A/G]CATAGTAGGCCAAGCGGCGGGCTTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PHLDB3 PubMed Links

Gene Details

Gene
PHLDB3
Gene Name
pleckstrin homology like domain family B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198850.3 2131 Missense Mutation GCG,GTG A567V NP_942147.3
XM_005259169.3 2131 Missense Mutation GCG,GTG A562V XP_005259226.1
XM_005259170.3 2131 Missense Mutation GCG,GTG A528V XP_005259227.1
XM_005259171.3 2131 Intron XP_005259228.1
XM_011527236.2 2131 Missense Mutation GCG,GTG A562V XP_011525538.1

View Full Product Details