Product Details

SNP ID
rs199971487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8522691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTGTTGTGCTCTGAGGGCGGA[C/T]GTGCCCGGGGTCGTCTGCTGGCTCC
Phenotype
MIM: 601480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO1F PubMed Links

Gene Details

Gene
MYO1F
Gene Name
myosin IF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012335.3 3059 Missense Mutation CAT,CGT H998R NP_036467.2
XM_011528024.1 3059 Missense Mutation CAT,CGT H1019R XP_011526326.1
XM_011528025.1 3059 Missense Mutation CAT,CGT H1015R XP_011526327.1
XM_011528026.1 3059 Missense Mutation CAT,CGT H990R XP_011526328.1
XM_011528027.1 3059 Missense Mutation CAT,CGT H974R XP_011526329.1
XM_011528028.2 3059 Intron XP_011526330.1
XM_017026821.1 3059 Missense Mutation CAT,CGT H994R XP_016882310.1
Gene
ZNF414
Gene Name
zinc finger protein 414
There are no transcripts associated with this gene.

View Full Product Details