Product Details

SNP ID
rs200386314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32692087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCGCCGCCGGCCGAGGGCTTC[C/T]GGCTGCTGCTGCTGCAGCGCTCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT19 PubMed Links

Gene Details

Gene
NUDT19
Gene Name
nudix hydrolase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105570.1 481 Missense Mutation CGG,TGG R43W NP_001099040.1
XM_017026806.1 481 Missense Mutation CGG,TGG R43W XP_016882295.1

View Full Product Details