Product Details

SNP ID
rs199585523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10514357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGCGCCGCGCGTTGGCGCGTA[C/T]CTGGCAGTAGATGCGCGCGTAGAGT
Phenotype
MIM: 605146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S1PR5 PubMed Links

Gene Details

Gene
S1PR5
Gene Name
sphingosine-1-phosphate receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166215.1 842 Missense Mutation ATA,GTA I219V NP_001159687.1
NM_030760.4 842 Missense Mutation ATA,GTA I219V NP_110387.1

View Full Product Details