Product Details

SNP ID
rs199692622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5724853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGTGTTTAATCTGATACAGGA[C/T]GTTCAAGGGGTATGTGGCTCCATGC
Phenotype
MIM: 605490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATSPERD PubMed Links

Gene Details

Gene
CATSPERD
Gene Name
cation channel sperm associated auxiliary subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152784.3 470 Silent Mutation GAC,GAT D39D NP_689997.3
XM_011527885.2 470 Silent Mutation GAC,GAT D39D XP_011526187.1
XM_011527886.2 470 Silent Mutation GAC,GAT D39D XP_011526188.1
XM_011527887.2 470 Silent Mutation GAC,GAT D39D XP_011526189.1
XM_011527888.2 470 Silent Mutation GAC,GAT D39D XP_011526190.1
XM_011527889.2 470 Silent Mutation GAC,GAT D39D XP_011526191.1
XM_011527890.2 470 Silent Mutation GAC,GAT D39D XP_011526192.1
XM_011527891.1 470 Intron XP_011526193.1
XM_011527892.1 470 Intron XP_011526194.1
XM_017026564.1 470 Silent Mutation GAC,GAT D39D XP_016882053.1
XM_017026565.1 470 Intron XP_016882054.1
XM_017026566.1 470 Silent Mutation GAC,GAT D39D XP_016882055.1
Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details