Product Details

SNP ID
rs199903417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16164737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACCTGAGGAAGTCTGGTGCCC[A/G]GAGGATCATGTTCTGGAAATCTTCC
Phenotype
MIM: 610645 MIM: 608349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIB3 PubMed Links

Gene Details

Gene
CIB3
Gene Name
calcium and integrin binding family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300922.1 573 Missense Mutation CGG,TGG R126W NP_001287851.1
NM_054113.3 573 Missense Mutation CGG,TGG R175W NP_473454.1
Gene
HSH2D
Gene Name
hematopoietic SH2 domain containing
There are no transcripts associated with this gene.

View Full Product Details