Product Details

SNP ID
rs193920819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35270514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAGGCTGTCAGCGGGGAGGCAC[A/G]ATTTGCCTATTTCCCAGCGTCCAGT
Phenotype
MIM: 616582 MIM: 600390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSR PubMed Links

Gene Details

Gene
LSR
Gene Name
lipolysis stimulated lipoprotein receptor
There are no transcripts associated with this gene.

Gene
USF2
Gene Name
upstream transcription factor 2, c-fos interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321150.1 520 Intron NP_001308079.1
NM_003367.3 520 Missense Mutation CAA,CGA Q166R NP_003358.1
NM_207291.2 520 Missense Mutation CAA,CGA Q99R NP_997174.1
XM_005259197.3 520 Missense Mutation CAA,CGA Q166R XP_005259254.1
XM_011527260.2 520 Missense Mutation CAA,CGA Q166R XP_011525562.1
XM_011527261.2 520 Missense Mutation CAA,CGA Q166R XP_011525563.1
XM_017027199.1 520 Missense Mutation CAA,CGA Q99R XP_016882688.1

View Full Product Details