Product Details
- SNP ID
-
rs199764677
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58433326 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCACTTCCATAAGGCTCCACTCA[A/G]GTATGAATCTTTTTATGCTGTGCAA
- Phenotype
-
MIM: 604074
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF132
PubMed Links
Gene Details
- Gene
- ZNF132
- Gene Name
- zinc finger protein 132
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003433.3 |
2519 |
Silent Mutation |
ACC,ACT |
T706T |
NP_003424.3 |
View Full Product Details