Product Details

SNP ID
rs199766242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36891806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCACTATTAAAGGCCTTCCCAC[A/C]CTGCTTACATTCATAAGGTTTCTCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF345 PubMed Links

Gene Details

Gene
ZNF345
Gene Name
zinc finger protein 345
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242472.1 1350 Intron NP_001229401.1
NM_001242474.1 1350 Intron NP_001229403.1
NM_001242475.1 1350 Intron NP_001229404.1
NM_001242476.1 1350 Intron NP_001229405.1
NM_003419.4 1350 Intron NP_003410.1
XM_011526704.2 1350 Intron XP_011525006.1
XM_017026568.1 1350 Intron XP_016882057.1
XM_017026569.1 1350 Intron XP_016882058.1
XM_017026570.1 1350 Intron XP_016882059.1
XM_017026571.1 1350 Intron XP_016882060.1
XM_017026572.1 1350 Intron XP_016882061.1
XM_017026573.1 1350 Intron XP_016882062.1
Gene
ZNF829
Gene Name
zinc finger protein 829
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037232.3 1350 Missense Mutation GGT,TGT G329C NP_001032309.2
NM_001171979.1 1350 Missense Mutation GGT,TGT G410C NP_001165450.1
XM_005258876.4 1350 Missense Mutation GGT,TGT G329C XP_005258933.1
XM_011526933.2 1350 Missense Mutation GGT,TGT G329C XP_011525235.1

View Full Product Details