Product Details

SNP ID
rs199872561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11722710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCACATTGTGTACATTTATAG[A/G]GTTTCTCTCCGGTGTGAGTTCTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 887 Missense Mutation NP_001073962.1
NM_001297610.1 887 Missense Mutation NP_001284539.1
NM_017507.1 887 Missense Mutation NP_059977.1
XM_017026941.1 887 Missense Mutation XP_016882430.1
XM_017026942.1 887 Missense Mutation XP_016882431.1

View Full Product Details