Product Details

SNP ID
rs199874894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51643962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGAGGCAGGGGGGTTGCTGT[A/C]AACTGTGCAGGCGAGGAACAGGGAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SIGLEC14 PubMed Links

Gene Details

Gene
SIGLEC14
Gene Name
sialic acid binding Ig like lectin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098612.1 949 Missense Mutation GAC,TAC D277Y NP_001092082.1
XM_017026113.1 949 Intron XP_016881602.1

View Full Product Details