Product Details

SNP ID
rs56186508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54803892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATCATGATCTTTCTTTGCAGG[A/G]TTCTTCTTGGACCAGAGTGTGTGGG
Phenotype
MIM: 604945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIR2DL4 PubMed Links

Gene Details

Gene
KIR2DL4
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080770.1 82 Silent Mutation GGA,GGG G14G NP_001074239.1
NM_001080772.1 82 Silent Mutation GGA,GGG G14G NP_001074241.1
NM_002255.5 82 Silent Mutation GGA,GGG G14G NP_002246.5
Gene
LOC102724300
Gene Name
uncharacterized LOC102724300
There are no transcripts associated with this gene.

View Full Product Details