Product Details
- SNP ID
-
rs200185005
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:52613377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCACATTTGTAAGGTTTCTCTCC[A/G]GTATGGATTCTGTGATGTTGTACAA
- Phenotype
-
MIM: 194558
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107983997
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11545619,rs568637119] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107983997
- Gene Name
- zinc finger protein 816-like
There are no transcripts associated with this gene.
- Gene
- ZNF701
- Gene Name
- zinc finger protein 701
- Gene
- ZNF83
- Gene Name
- zinc finger protein 83
View Full Product Details