Product Details

SNP ID
rs200153057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35615256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCATGGAGCAGGCACGTCAGCAC[A/T]CTCAAGACACCCAGCGTCGAGCTCT
Phenotype
MIM: 613432
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HAUS5 PubMed Links

Gene Details

Gene
HAUS5
Gene Name
HAUS augmin like complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015302.1 293 Missense Mutation ACT,TCT T119S NP_056117.1
XM_011526684.2 293 Missense Mutation ACT,TCT T19S XP_011524986.1

View Full Product Details