Product Details

SNP ID
rs199665148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57572780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATATGGCTTTTCTCCTGTGTGA[C/G]TTCTCTGGTGTCGAACAAGAGTGGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF416 PubMed Links

Gene Details

Gene
ZNF416
Gene Name
zinc finger protein 416
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017879.1 1295 Missense Mutation ACT,AGT T375S NP_060349.1
XM_005259051.3 1295 Missense Mutation ACT,AGT T303S XP_005259108.1

View Full Product Details