Product Details

SNP ID
rs199807704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9619454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCACGGGACAAATAAATGGCTGC[C/T]ATTCTCTGAAGCTGATGGTGTGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF561 PubMed Links

Gene Details

Gene
ZNF561
Gene Name
zinc finger protein 561
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152289.2 176 Missense Mutation ATA,ATG I1M NP_689502.2
XM_005260150.2 176 Missense Mutation ATA,ATG I1M XP_005260207.1
XM_005260151.2 176 UTR 5 XP_005260208.1
XM_006722938.2 176 Missense Mutation AGC,GGC S12G XP_006723001.1
XM_011528433.2 176 Missense Mutation AGC,GGC S12G XP_011526735.1
XM_011528434.2 176 Missense Mutation AGC,GGC S12G XP_011526736.1
XM_011528436.2 176 UTR 5 XP_011526738.1
XM_011528437.2 176 UTR 5 XP_011526739.1
XM_017027478.1 176 UTR 5 XP_016882967.1
XM_017027479.1 176 UTR 5 XP_016882968.1
XM_017027480.1 176 UTR 5 XP_016882969.1
XM_017027481.1 176 UTR 5 XP_016882970.1
XM_017027482.1 176 UTR 5 XP_016882971.1
Gene
ZNF561-AS1
Gene Name
ZNF561 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details