Product Details
- SNP ID
-
rs200227335
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55440695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGGACCGCCGATCCCCGGGGTCA[C/G]GGAGCTGCTTCGGGCCCGGTCCGGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA7
PubMed Links
Gene Details
- Gene
- SHISA7
- Gene Name
- shisa family member 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145176.1 |
742 |
Missense Mutation |
CTG,GTG |
L248V |
NP_001138648.1 |
View Full Product Details