Product Details
- SNP ID
-
rs199669425
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55316918 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAGAAGTAGACGTTACCCAAGA[C/T]GAAGGCAAGGAAGGCCCCTGCCAAG
- Phenotype
-
MIM: 609235
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BRSK1
PubMed Links
Gene Details
- Gene
- BRSK1
- Gene Name
- BR serine/threonine kinase 1
There are no transcripts associated with this gene.
- Gene
- TMEM150B
- Gene Name
- transmembrane protein 150B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001085488.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
NP_001078957.1 |
NM_001282011.1 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
NP_001268940.1 |
XM_005258812.3 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_005258869.1 |
XM_011526850.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525152.1 |
XM_011526851.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525153.1 |
XM_011526852.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525154.1 |
XM_011526853.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525155.1 |
XM_011526854.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525156.1 |
XM_011526855.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525157.1 |
XM_011526856.2 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_011525158.1 |
XM_011526857.2 |
754 |
Missense Mutation |
ATC,GTC |
I65V |
XP_011525159.1 |
XM_017026667.1 |
754 |
Missense Mutation |
ATC,GTC |
I125V |
XP_016882156.1 |
XM_017026668.1 |
754 |
Missense Mutation |
ATC,GTC |
I65V |
XP_016882157.1 |
View Full Product Details