Product Details

SNP ID
rs202027527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:841068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGCTGCTGGCCTTGCTGCTGAG[C/T]GGTGAGTGAGCCACGTGCCCATCCA
Phenotype
MIM: 162815 MIM: 130130 MIM: 177020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AZU1 PubMed Links

Gene Details

Gene
AZU1
Gene Name
azurocidin 1
There are no transcripts associated with this gene.

Gene
ELANE
Gene Name
elastase, neutrophil expressed
There are no transcripts associated with this gene.

Gene
PRTN3
Gene Name
proteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002777.3 104 Silent Mutation AGC,AGT S20S NP_002768.3
XM_011528136.1 104 Silent Mutation AGC,AGT S20S XP_011526438.1

View Full Product Details