Product Details

SNP ID
rs200884173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13913270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGCCAGACAAGCTGGAGACAGC[A/G]CCAAGATGCGGCGCTACGATCGGGG
Phenotype
MIM: 610055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf57 PubMed Links

Gene Details

Gene
C19orf57
Gene Name
chromosome 19 open reading frame 57
There are no transcripts associated with this gene.

Gene
CC2D1A
Gene Name
coiled-coil and C2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017721.4 780 Missense Mutation ACC,GCC T161A NP_060191.3
XM_005259972.2 780 Missense Mutation ACC,GCC T161A XP_005260029.1
XM_005259973.2 780 Missense Mutation ACC,GCC T161A XP_005260030.1
XM_005259974.2 780 Missense Mutation ACC,GCC T161A XP_005260031.1
XM_005259975.2 780 Missense Mutation ACC,GCC T161A XP_005260032.1

View Full Product Details