Product Details

SNP ID
rs200884858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51612344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGCATAATGGAGCTCCTTTT[A/G]TTCTTCCAAGGGAGGGGCATCCCCA
Phenotype
MIM: 604200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC5 PubMed Links

Gene Details

Gene
SIGLEC5
Gene Name
sialic acid binding Ig like lectin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003830.3 1682 Nonsense Mutation CAA,TAA Q515* NP_003821.1
XM_011527438.2 1682 Silent Mutation AAC,AAT N487N XP_011525740.1
XM_017027419.1 1682 Nonsense Mutation CAA,TAA Q524* XP_016882908.1

View Full Product Details