Product Details

SNP ID
rs201436589
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49489871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTCCTGGTGCTTGATGGTCGA[A/G]GCCATCTCCTGGGCCGCCTGGCGGC
Phenotype
MIM: 600007 MIM: 180471
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FLT3LG PubMed Links
Additional Information
For this assay, SNP(s) [rs139354594] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FLT3LG
Gene Name
fms related tyrosine kinase 3 ligand
There are no transcripts associated with this gene.

Gene
RPL13A
Gene Name
ribosomal protein L13a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270491.1 Intron NP_001257420.1
NM_012423.3 Intron NP_036555.1
Gene
RPS11
Gene Name
ribosomal protein S11
There are no transcripts associated with this gene.

Gene
SNORD32A
Gene Name
small nucleolar RNA, C/D box 32A
There are no transcripts associated with this gene.

Gene
SNORD33
Gene Name
small nucleolar RNA, C/D box 33
There are no transcripts associated with this gene.

Gene
SNORD34
Gene Name
small nucleolar RNA, C/D box 34
There are no transcripts associated with this gene.

Gene
SNORD35A
Gene Name
small nucleolar RNA, C/D box 35A
There are no transcripts associated with this gene.

Gene
SNORD35B
Gene Name
small nucleolar RNA, C/D box 35B
There are no transcripts associated with this gene.

View Full Product Details