Product Details

SNP ID
rs201436617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37885153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACAGGCAGCAGAACAGCCGTT[A/C]CCCGGGAACTAGCTCTGTGGCTTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WDR87 PubMed Links

Gene Details

Gene
WDR87
Gene Name
WD repeat domain 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291088.1 8772 Missense Mutation NP_001278017.1
NM_031951.4 8772 Missense Mutation NP_114157.4
XM_011527360.2 8772 Nonsense Mutation XP_011525662.1

View Full Product Details