Product Details

SNP ID
rs201887538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49513407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGTCGTCCTCTCAGCATGGGG[A/G]TCCCGCGGCCTCAGCCCTGGGCGCT
Phenotype
MIM: 601437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCGRT PubMed Links

Gene Details

Gene
FCGRT
Gene Name
Fc fragment of IgG receptor and transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136019.2 229 Missense Mutation ATC,GTC I3V NP_001129491.1
NM_004107.4 229 Missense Mutation ATC,GTC I3V NP_004098.1
XM_005258657.1 229 Intron XP_005258714.1
XM_011526627.2 229 Missense Mutation ATC,GTC I3V XP_011524929.1

View Full Product Details