Product Details
- SNP ID
-
rs202170300
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:54218831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCAGGACGCTGGAAATCAGTCTT[A/T]CTCTGGTCTGGGTGAAGATGGACAG
- Phenotype
-
MIM: 604820
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LILRB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3852891,rs3852892] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LILRB3
- Gene Name
- leukocyte immunoglobulin like receptor B3
View Full Product Details