Product Details

SNP ID
rs202202193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51890741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGCTGTGACTTGCTGTGGAAGG[A/G]TTTTCCACCTTCACTGAATCCCCCC
Phenotype
MIM: 611903
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF577 PubMed Links

Gene Details

Gene
ZNF577
Gene Name
zinc finger protein 577
There are no transcripts associated with this gene.

Gene
ZNF649
Gene Name
zinc finger protein 649
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023074.3 1721 Silent Mutation AAC,AAT N465N NP_075562.2
Gene
ZNF649-AS1
Gene Name
ZNF649 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details