Product Details

SNP ID
rs200576016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14472616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGCAGCGAGCTGGCCTCCCAGG[A/C]GCTCGGTGTTAGGCCCAGCCCCGCG
Phenotype
MIM: 605072 MIM: 601032 MIM: 176802
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GIPC1 PubMed Links

Gene Details

Gene
GIPC1
Gene Name
GIPC PDZ domain containing family member 1
There are no transcripts associated with this gene.

Gene
PKN1
Gene Name
protein kinase N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002741.3 1271 Intron NP_002732.3
NM_213560.1 1271 Intron NP_998725.1
XM_011528128.1 1271 Intron XP_011526430.1
Gene
PTGER1
Gene Name
prostaglandin E receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000955.2 1271 Missense Mutation GCC,TCC A385S NP_000946.2

View Full Product Details